Anti-6x His Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: AD1.1.10]
Lieferant: Agrisera
AS152931
AGRIAS152931EA 482 EUR
AGRIAS152931
Anti-6x His Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: AD1.1.10]
Antikörper
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag.,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag.
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: SureLight® RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana