Together with gamma - glutamyl Cys sythetase (γ-ECS) and glutathione synthetase, phytochelatin synthetase (PS), is an enzyme involved in, and required for, synthetizing phytochelatins (PC).
- Recommended dilution - 1:1000 (ELISA), (WB)
In certain plants, deficiency of such enzymes leads to low tolerance to metal contamination while mutants over-expressing them are highly tolerant to metal contamination and may be used in phyto-remediation of heavy metals.
Type:
Antigen: PS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Schizosaccharomyces pombe