Anti-GST-pi Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences
ADI-MSA-102-E ADIMSA102E
ALEXADI-MSA-102-EEA 0 EUR
ALEXADI-MSA-102-E ENZOADIMSA102E
Anti-GST-pi Rabbit Polyclonal Antibody
Antikörper
The GST superfamily of enzymes play an important role in detoxification of xenobiotics by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione.

  • Dilution range: Immunohistochemistry (1:500 - 1:1,000), Western blot (1:500 - 1:1,000)
  • Detects a band of ~26kDa by western blot

Type: Primary
Antigen: GST-P, Glutathione S-transferase P, GSTP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR