Anti-Rad51C Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A100120-100
ANTIA100120-100EA 475 EUR
ANTIA100120-100
Anti-Rad51C Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to RAD51L2 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Type: Primary
Antigen: Rad51C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR