Anti-OR4L1 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A100899-100
ANTIA100899-100EA 475 EUR
ANTIA100899-100
Anti-OR4L1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to OR4L1 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: OR4L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR