Anti-ADH7 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A101562-100
ANTIA101562-100EA 475 EUR
ANTIA101562-100
Anti-ADH7 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to ADH7 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen: ADH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR