Anti-Glutamyl hydrolase gamma Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A101627-100
ANTIA101627-100EA 475 EUR
ANTIA101627-100
Anti-Glutamyl hydrolase gamma Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to GGH for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Glutamyl hydrolase gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR