Anti-RRM1 Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A101693-100
ANTIA101693-100EA 460 EUR
ANTIA101693-100
Anti-RRM1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to RRM1 for WB and IF with samples derived from human, mouse, rat, hamster and xenopus.

  • Validated Applications: WB, IF
  • Recommended Dilutions: WB: 1:1,000 to 1:2,000, IF: 1:100

Type: Primary
Antigen: RRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Xenopus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR