Anti-Rad51C Rabbit Polyclonal Antibody
Lieferant: ANTIBODIES.COM
A12048-100
ANTIA12048-100EA 415 EUR
ANTIA12048-100
Anti-Rad51C Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Rad51C for WB with samples derived from human and mouse.
  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: Rad51C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse