Anti-CD15 Mouse Monoclonal Antibody [clone: MMA] (PerCP-Cyanine 5.5)
Lieferant: ANTIBODIES.COM
A121929-100
ANTIA121929-100EA 485 EUR
ANTIA121929-100
Anti-CD15 Mouse Monoclonal Antibody [clone: MMA] (PerCP-Cyanine 5.5)
Antikörper
Mouse monoclonal [MMA] antibody to CD15 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MMA
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human