Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7] (PerCP-Cyanine 5.5)
Lieferant: ANTIBODIES.COM
A121932-100
ANTIA121932-100EA 485 EUR
ANTIA121932-100
Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7] (PerCP-Cyanine 5.5)
Antikörper
Mouse monoclonal [4G7] antibody to CD19 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human