Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (PerCP-Cyanine 5.5)
Lieferant: Antibodies.com
A121944-100
ANTIA121944-100EA 485 EUR
ANTIA121944-100
Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (PerCP-Cyanine 5.5)
Antikörper
Mouse monoclonal [OKT3] antibody to CD3 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human