Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (PerCP-Cyanine 5.5)

Lieferant: ANTIBODIES.COM
A121954-100
ANTIA121954-100EA 485 EUR
ANTIA121954-100
Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (PerCP-Cyanine 5.5)
Antikörper
Mouse monoclonal [2D1] antibody to CD45 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2D1
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR