Anti-CD64 Mouse Monoclonal Antibody [clone: 10.1] (PerCP-Cyanine 5.5)
Lieferant: Antibodies.com
A121961-100
ANTIA121961-100EA 485 EUR
ANTIA121961-100
Anti-CD64 Mouse Monoclonal Antibody [clone: 10.1] (PerCP-Cyanine 5.5)
Antikörper
Mouse monoclonal [10.1] antibody to CD64 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human and Non-Human Primates.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 10.1
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates