Anti-CD11c Mouse Monoclonal Antibody [clone: BU15] (PE-Cyanine 7)

Lieferant: ANTIBODIES.COM
A121978-100
ANTIA121978-100EA 375 EUR
ANTIA121978-100
Anti-CD11c Mouse Monoclonal Antibody [clone: BU15] (PE-Cyanine 7)
Antikörper
Mouse monoclonal [BU15] antibody to CD11c (PE-Cyanine 7) for Flow Cytometry with samples derived from human and monkey.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: BU15
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR