Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5] (PE-Cyanine 7)

Lieferant: Antibodies.com
A121987-100
ANTIA121987-100EA 375 EUR
ANTIA121987-100
Anti-CXCR4 Mouse Monoclonal Antibody [clone: 12G5] (PE-Cyanine 7)
Antikörper
Mouse monoclonal [12G5] antibody to CXCR4 (PE-Cyanine 7) for Flow Cytometry with samples derived from human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Non-Human Primates
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR