Anti-CD19 Mouse Monoclonal Antibody [clone: LT19] (PE-Cyanine 7)
Lieferant: ANTIBODIES.COM
A121989-100
ANTIA121989-100EA 375 EUR
ANTIA121989-100
Anti-CD19 Mouse Monoclonal Antibody [clone: LT19] (PE-Cyanine 7)
Antikörper
Mouse monoclonal [LT19] antibody to CD19 (PE-Cyanine 7) for Flow Cytometry with samples derived from human.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: LT19
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human