Anti-CD21 Mouse Monoclonal Antibody [clone: LT21] (PE-Cyanine 7)

Lieferant: ANTIBODIES.COM
A121995-100
ANTIA121995-100EA 375 EUR
ANTIA121995-100
Anti-CD21 Mouse Monoclonal Antibody [clone: LT21] (PE-Cyanine 7)
Antikörper
Mouse monoclonal [LT21] antibody to CD21 (PE-Cyanine 7) for Flow Cytometry with samples derived from Porcine, Human, Canine and Bovine.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: LT21
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Porcine;Human;Dog;Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR