Anti-CD22 Mouse Monoclonal Antibody [clone: IS7] (PE-Cyanine 7)

Lieferant: ANTIBODIES.COM
A121996-100
ANTIA121996-100EA 375 EUR
ANTIA121996-100
Anti-CD22 Mouse Monoclonal Antibody [clone: IS7] (PE-Cyanine 7)
Antikörper
Mouse monoclonal [IS7] antibody to CD22 (PE-Cyanine 7) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: IS7
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR