Anti-EGFR Mouse Monoclonal Antibody [clone: EGFR1] (PE-Cyanine 7)

Lieferant: Antibodies.com
A122032-100
ANTIA122032-100EA 375 EUR
ANTIA122032-100
Anti-EGFR Mouse Monoclonal Antibody [clone: EGFR1] (PE-Cyanine 7)
Antikörper
Mouse monoclonal [EGFR1] antibody to EGFR (PE-Cyanine 7) for Flow Cytometry with samples derived from human and Horse.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: EGFR1
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Horse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR