Anti-CD117 Mouse Monoclonal Antibody [clone: 104D2] (PE-Cyanine 5)
Lieferant: ANTIBODIES.COM
A122041-100
ANTIA122041-100EA 345 EUR
ANTIA122041-100
Anti-CD117 Mouse Monoclonal Antibody [clone: 104D2] (PE-Cyanine 5)
Antikörper
Mouse monoclonal [104D2] antibody to CD117 (PE-Cyanine 5) for Flow Cytometry with samples derived from human, Bovine and Non-Human Primates.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: 104D2
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Bovine, Non-Human Primates