Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 5)

Lieferant: ANTIBODIES.COM
A122062-100
ANTIA122062-100EA 345 EUR
ANTIA122062-100
Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 5)
Antikörper
Mouse monoclonal [MEM-28] antibody to CD45 (PE-Cyanine 5) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: MEM-28
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR