Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE-Cyanine 5)

Lieferant: ANTIBODIES.COM
A122064-100
ANTIA122064-100EA 345 EUR
ANTIA122064-100
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE-Cyanine 5)
Antikörper
Mouse monoclonal [L17F12] antibody to CD5 (PE-Cyanine 5) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR