Anti-CD62P Mouse Monoclonal Antibody [clone: AK4] (PE-Cyanine 5)

Lieferant: Antibodies.com
A122066-100
ANTIA122066-100EA 345 EUR
ANTIA122066-100
Anti-CD62P Mouse Monoclonal Antibody [clone: AK4] (PE-Cyanine 5)
Antikörper
Mouse monoclonal [AK4] antibody to CD62P (PE-Cyanine 5) for Flow Cytometry with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD62P
Clonality: Monoclonal
Clone: AK4
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR