Anti-CD86 Mouse Monoclonal Antibody [clone: BU63] (PE-Cyanine 5)

Lieferant: Antibodies.com
A122074-100
ANTIA122074-100EA 345 EUR
ANTIA122074-100
Anti-CD86 Mouse Monoclonal Antibody [clone: BU63] (PE-Cyanine 5)
Antikörper
Mouse monoclonal [BU63] antibody to CD86 (PE-Cyanine 5) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: BU63
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR