Anti-HLA DR Mouse Monoclonal Antibody [clone: L243] (APC (Allophycocyanin)-Cy7®)

Lieferant: ANTIBODIES.COM
A122111-100
ANTIA122111-100EA 375 EUR
ANTIA122111-100
Anti-HLA DR Mouse Monoclonal Antibody [clone: L243] (APC (Allophycocyanin)-Cy7®)
Antikörper
Mouse monoclonal [L243] antibody to HLA DR (APC-Cyanine 7) for Flow Cytometry with samples derived from human, Canine and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: HLA DR
Clonality: Monoclonal
Clone: L243
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Dog, Non-Human Primates
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR