Anti-Tec Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A12533-100
ANTIA12533-100EA 415 EUR
ANTIA12533-100
Anti-Tec Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Tec for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: Tec
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR