Anti-TGF alpha Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A12614-100
ANTIA12614-100EA 415 EUR
ANTIA12614-100
Anti-TGF alpha Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to TGF alpha for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:100 to 1:50

Type: Primary
Antigen: TGF alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR