Anti-IgG H&L Goat Polyclonal Antibody (Cyanine 3)
Lieferant: ANTIBODIES.COM
A17341-100
ANTIA17341-100EA 130 EUR
ANTIA17341-100
Anti-IgG H&L Goat Polyclonal Antibody (Cyanine 3)
Antikörper
Goat Anti-Rabbit IgG H&L Antibody (Cyanine 3) for ICC/IF and Flow Cytometry with samples derived from rabbit.
  • Validated applications: ICC/IF, flow cytometry
  • Recommended dilutions: ICC/IF: 1:100 to 1:800, flow cytometry: 1:100 to 1:80
Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Cyanine 3
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit