Anti-IgG H&L Goat Polyclonal Antibody (Cyanine 3)

Lieferant: ANTIBODIES.COM
A17349-100
ANTIA17349-100EA 130 EUR
ANTIA17349-100
Anti-IgG H&L Goat Polyclonal Antibody (Cyanine 3)
Antikörper
Goat Anti-Mouse IgG H&L Antibody (Cyanine 3) for ICC/IF and Flow Cytometry with samples derived from mouse.

  • Validated applications: ICC/IF, flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100 to 1:800, ICC/IF: 1:50 to 1:20

Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Cyanine 3
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR