Anti-Albumin Mouse Monoclonal Antibody [clone: ALB/2356]

Lieferant: ANTIBODIES.COM
A248502-100
ANTIA248502-100EA 520 EUR
ANTIA248502-100
Anti-Albumin Mouse Monoclonal Antibody [clone: ALB/2356]
Antikörper
Mouse monoclonal [ALB/2356] antibody to Albumin for ELISA, flow cytometry, IF and WB with samples derived from human.

  • Validated applications: ELISA, Flow cytometry, IF, WB
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 2 to 4 µg/ml, WB: 1 to 2 µg/ml

Type: Primary
Antigen: Albumin
Clonality: Monoclonal
Clone: ALB/2356
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR