Anti-Lambda Light Chain Mouse Recombinant Antibody [clone: rLLC/1738]
Lieferant: ANTIBODIES.COM
A248999-100
ANTIA248999-100EA 520 EUR
ANTIA248999-100
Anti-Lambda Light Chain Mouse Recombinant Antibody [clone: rLLC/1738]
Antikörper
Recombinant mouse monoclonal [rLLC/1738] antibody to Lambda Light Chain for IHC-P with samples derived from human.
  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Lambda Light Chain
Clonality: Recombinant
Clone: rLLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human