Anti-IL2 Receptor alpha Mouse Monoclonal Antibody [clone: IL2RA/2395]

Lieferant: ANTIBODIES.COM
A249018-100
ANTIA249018-100EA 520 EUR
ANTIA249018-100
Anti-IL2 Receptor alpha Mouse Monoclonal Antibody [clone: IL2RA/2395]
Antikörper
Mouse monoclonal [IL2RA/2395] antibody to IL2 Receptor alpha for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 2 to 4 µg/ml

Type: Primary
Antigen: IL2 Receptor alpha
Clonality: Monoclonal
Clone: IL2RA/2395
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR