Anti-CD43 Rabbit Recombinant Antibody [clone: SPN/2049R]

Lieferant: ANTIBODIES.COM
A250033-100
ANTIA250033-100EA 520 EUR
ANTIA250033-100
Anti-CD43 Rabbit Recombinant Antibody [clone: SPN/2049R]
Antikörper
Recombinant rabbit monoclonal [SPN/2049R] antibody to CD43 for WB, flow cytometry, IF and IHC-P with samples derived from human.

  • Validated applications: WB, Flow cytometry, IF, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD43
Clonality: Recombinant
Clone: SPN/2049R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR