Anti-NAPSIN A Mouse Monoclonal Antibody [clone: NAPSA/3306]

Lieferant: ANTIBODIES.COM
A250672-100
ANTIA250672-100EA 520 EUR
ANTIA250672-100
Anti-NAPSIN A Mouse Monoclonal Antibody [clone: NAPSA/3306]
Antikörper
Mouse monoclonal [NAPSA/3306] antibody to NAPSIN A for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: NAPSIN A
Clonality: Monoclonal
Clone: NAPSA/3306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR