Anti-Podocalyxin Rabbit Recombinant Antibody [clone: PDPN/4009R]

Lieferant: ANTIBODIES.COM
A251332-100
ANTIA251332-100EA 520 EUR
ANTIA251332-100
Anti-Podocalyxin Rabbit Recombinant Antibody [clone: PDPN/4009R]
Antikörper
Recombinant rabbit monoclonal [PDPN/4009R] antibody to Podocalyxin for ELISA, IF and IHC-P with samples derived from Human.

  • Validated applications: ELISA, IF, IHC-P
  • Recommended dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Podocalyxin
Clonality: Recombinant
Clone: PDPN/4009R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR