Anti-EPO Mouse Monoclonal Antibody [clone: EPO/1368]

Lieferant: ANTIBODIES.COM
A251622-100
ANTIA251622-100EA 520 EUR
ANTIA251622-100
Anti-EPO Mouse Monoclonal Antibody [clone: EPO/1368]
Antikörper
Mouse monoclonal [EPO/1368] antibody to EPO for ELISA, WB and IHC-P with samples derived from Human.

  • Validated applications: ELISA, WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 2 to 4 µg/ml

Type: Primary
Antigen: EPO
Clonality: Monoclonal
Clone: EPO/1368
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR