Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]

Lieferant: ANTIBODIES.COM
A252311-100
ANTIA252311-100EA 520 EUR
ANTIA252311-100
Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]
Antikörper
Recombinant mouse monoclonal [rKRT6A/2100] antibody to Cytokeratin 6A for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 6A
Clonality: Recombinant
Clone: rKRT6A/2100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR