Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/520]

Lieferant: Antibodies.com
A252579-100
ANTIA252579-100EA 520 EUR
ANTIA252579-100
Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/520]
Antikörper
Mouse monoclonal [MUC1/520] antibody to MUC1 for IHC with samples derived from Human.

  • Validated applications: IHC
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: MUC1/520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR