Anti-Bcl-2 Rabbit Recombinant Antibody [clone: BCL2/2210R]

Lieferant: ANTIBODIES.COM
A253054-100
ANTIA253054-100EA 520 EUR
ANTIA253054-100
Anti-Bcl-2 Rabbit Recombinant Antibody [clone: BCL2/2210R]
Antikörper
Recombinant rabbit monoclonal [BCL2/2210R] antibody to Bcl-2 for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Bcl-2
Clonality: Recombinant
Clone: BCL2/2210R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR