Anti-C3d Mouse Monoclonal Antibody [clone: C3D/2891]

Lieferant: ANTIBODIES.COM
A253394-100
ANTIA253394-100EA 520 EUR
ANTIA253394-100
Anti-C3d Mouse Monoclonal Antibody [clone: C3D/2891]
Antikörper
Mouse monoclonal [C3D/2891] antibody to C3d for ELISA with samples derived from human.

  • Validated Applications: ELISA

Type: Primary
Antigen: C3d
Clonality: Monoclonal
Clone: C3D/2891
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR