Anti-NAPSIN A Mouse Monoclonal Antibody [Clone: NAPSA/3306]
Lieferant: ANTIBODIES.COM
A253852-100
ANTIA253852-100EA 520 EUR
ANTIA253852-100
Anti-NAPSIN A Mouse Monoclonal Antibody [Clone: NAPSA/3306]
Antikörper
Mouse monoclonal [NAPSA/3306] antibody to NAPSIN A for IHC-P with samples derived from human.
  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: NAPSIN A
Clonality: Monoclonal
Clone: NAPSA/3306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human