Anti-NuMA Mouse Monoclonal Antibody [Clone: A73-B/D12]
Lieferant: ANTIBODIES.COM
A254329-100
ANTIA254329-100EA 520 EUR
ANTIA254329-100
Anti-NuMA Mouse Monoclonal Antibody [Clone: A73-B/D12]
Antikörper
Mouse monoclonal [A73-B/D12] antibody to NuMA for Flow Cytometry, IF and IHC-P with samples derived from human.
  • Validated Applications: Flow Cytometry, IF, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: NuMA
Clonality: Monoclonal
Clone: A73-B/D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human