Anti-delta 1 Catenin Mouse Monoclonal Antibody [clone: CTNND1/4501]

Lieferant: ANTIBODIES.COM
A277567-100
ANTIA277567-100EA 520 EUR
ANTIA277567-100
Anti-delta 1 Catenin Mouse Monoclonal Antibody [clone: CTNND1/4501]
Antikörper
Mouse monoclonal [CTNND1/4501] antibody to delta 1 Catenin for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: delta 1 Catenin
Clonality: Monoclonal
Clone: CTNND1/4501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR