Anti-SPARC Mouse Monoclonal Antibody [Clone: OSTN/3933]

Lieferant: ANTIBODIES.COM
A277804-100
ANTIA277804-100EA 520 EUR
ANTIA277804-100
Anti-SPARC Mouse Monoclonal Antibody [Clone: OSTN/3933]
Antikörper
Mouse monoclonal [OSTN/3933] antibody to SPARC for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: OSTN/3933
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR