Anti-SPARC Mouse Monoclonal Antibody [Clone: OSTN/3755]
Lieferant: ANTIBODIES.COM
A277806-100
ANTIA277806-100EA 520 EUR
ANTIA277806-100
Anti-SPARC Mouse Monoclonal Antibody [Clone: OSTN/3755]
Antikörper
Mouse monoclonal [OSTN/3755] antibody to SPARC for WB and IHC-P with samples derived from human.
  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: OSTN/3755
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human