Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3758]

Lieferant: ANTIBODIES.COM
A277807-100
ANTIA277807-100EA 520 EUR
ANTIA277807-100
Anti-SPARC Mouse Monoclonal Antibody [clone: OSTN/3758]
Antikörper
Mouse monoclonal [OSTN/3758] antibody to SPARC for WB and IHC-P with samples derived from human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: OSTN/3758
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR