Anti-PU.1 Mouse Monoclonal Antibody [clone: PU1/2118]

Lieferant: ANTIBODIES.COM
A277809-100
ANTIA277809-100EA 520 EUR
ANTIA277809-100
Anti-PU.1 Mouse Monoclonal Antibody [clone: PU1/2118]
Antikörper
Mouse monoclonal [PU1/2118] antibody to PU.1 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: PU.1
Clonality: Monoclonal
Clone: PU1/2118
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR