Anti-HLA DP + DQ + DR Mouse Monoclonal Antibody [clone: HLA-Pan/6663]

Lieferant: ANTIBODIES.COM
A277868-100
ANTIA277868-100EA 520 EUR
ANTIA277868-100
Anti-HLA DP + DQ + DR Mouse Monoclonal Antibody [clone: HLA-Pan/6663]
Antikörper
Mouse monoclonal [HLA-Pan/6663] antibody to HLA DP + DQ + DR for Flow Cytometry and IHC-P with samples derived from human.

  • Validated Applications: Flow Cytometry, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HLA DP + DQ + DR
Clonality: Monoclonal
Clone: HLA-Pan/6663
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR