Anti-AKR1B1 Rabbit Recombinant Antibody [clone: AKR1B1/7009R]

Lieferant: ANTIBODIES.COM
A277982-100
ANTIA277982-100EA 520 EUR
ANTIA277982-100
Anti-AKR1B1 Rabbit Recombinant Antibody [clone: AKR1B1/7009R]
Antikörper
Recombinant rabbit monoclonal [AKR1B1/7009R] antibody to AKR1B1 for WB and IHC-P with samples derived from human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: AKR1B1
Clonality: Recombinant
Clone: AKR1B1/7009R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR